Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012311.3 | NP_036443.1 | 13 | 5779 | ||
NR_045609.1 | 14 | 3354 | ||||
GenBank mRNA | BC037913.2 | - | 2 | 1927 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CK003350.1 | - | 1 | 145 | Chr. 10 - 7751049 - 7751721 [-] on Build GRCh38 |