Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006717015.3 | XP_006717078.1 | 7 | 4065 | ||
XM_011518392.2 | XP_011516694.1 | 8 | 4181 | |||
GenBank mRNA | AK096112.1 | - | 1 | 2683 |