Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPPL2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005254722.3 | XP_005254779.1 | 16 | 3665 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC036610.1 | - | 1 | 186 | Chr. 15 - 50704531 - 50706116 [-] on Build GRCh38 |