Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP4R3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK098321.1 | - | 1 | 266 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB259847.1 | - | 1 | 266 | Chr. 2 - 55550148 - 55550719 [-] on Build GRCh38 |