Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAZN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011541074.2 | XP_011539376.1 | 16 | 3209 | ||
XM_011541075.2 | XP_011539377.1 | 14 | 2865 | |||
XM_011541076.2 | XP_011539378.1 | 14 | 3010 | |||
XM_011541077.2 | XP_011539379.1 | 14 | 3989 | |||
XM_011541081.2 | XP_011539383.1 | 8 | 1689 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK095631.1 | - | 2 | 2829 | Chr. 1 - 15109946 - 15113444 [+] on Build GRCh38 |