Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAFIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046442.2 | 3 | 659 |
Gene Symbol: TEKT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005263876.2 | XP_005263933.1 | 1 | 247 | ||
XM_011510670.1 | XP_011508972.1 | 1 | 247 |
Gene Symbol: TEKT4P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038329.2 | 3 | 650 | |||
GenBank mRNA | DT217039.1 | - | 3 | 649 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR617898.1 | - | 1 | 528 | Chr. 2 - 94867486 - 94868761 [+] on Build GRCh38 |