Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ESRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BU934976.1 | - | 1 | 583 |
Gene Symbol: NFATC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004555.3 | NP_004546.1 | 11 | 6177 | ||
NM_173163.2 | NP_775186.1 | 11 | 6162 | |||
NM_173165.2 | NP_775188.1 | 10 | 6073 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR607695.1 | - | 1 | 1775 | Chr. 16 - 68227226 - 68229145 [+] on Build GRCh38 |