Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: C16orf72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011522462.2 | XP_011520764.1 | 4 | 4882 | ||
GenBank mRNA | AK057657.1 | - | 1 | 2246 | ||
AL832650.1 | - | 1 | 4711 |