Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STX8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011524079.2 | XP_011522381.1 | 1 | 390 | ||
GenBank mRNA | BC020924.2 | - | 1 | 395 |