Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: NFATC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136021.2 | NP_001129493.1 | 11 | 5303 | ||
NM_001258292.1 | NP_001245221.1 | 10 | 5215 | |||
NM_001258294.1 | NP_001245223.1 | 11 | 4789 | |||
NM_001258295.1 | NP_001245224.1 | 10 | 4701 | |||
NM_001258296.1 | NP_001245225.1 | 11 | 4867 | |||
NM_001258297.1 | NP_001245226.1 | 10 | 4779 | |||
NM_012340.4 | NP_036472.2 | 11 | 5381 | |||
NM_173091.3 | NP_775114.1 | 10 | 5293 | |||
XM_017027851.1 | XP_016883340.1 | 9 | 4594 | |||
GenBank mRNA | AK025758.1 | - | 1 | 1206 | ||
AK226146.1 | - | 10 | 5217 |