Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: NRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017005186.1 | XP_016860675.1 | 17 | 5360 | ||
GenBank mRNA | AF119893.2 | - | 1 | 829 | ||
AK024680.1 | - | 1 | 710 |