Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA1614-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK026359.1 | - | 2 | 1128 |
Gene Symbol: STX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011509058.1 | XP_011507360.1 | 8 | 2047 | ||
XM_011509059.1 | XP_011507361.1 | 7 | 1822 | |||
XM_017000006.1 | XP_016855495.1 | 6 | 1656 |