Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGF14-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036487.1 | 1 | 1091 | |||
GenBank mRNA | AF086059.1 | - | 1 | 327 | ||
AF339777.1 | - | 1 | 494 | |||
BC033993.1 | - | 1 | 1040 |