Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGF14-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036487.1 | 1 | 517 | |||
GenBank mRNA | BC033993.1 | - | 1 | 466 | ||
BX096285.1 | - | 1 | 517 |