Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLIPR1L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270396.1 | NP_001257325.1 | 6 | 2189 | ||
NR_072995.1 | 7 | 2319 | ||||
XM_011537949.2 | XP_011536251.1 | 6 | 2103 | |||
XM_011537950.2 | XP_011536252.1 | 6 | 2075 | |||
XM_017018848.1 | XP_016874337.1 | 7 | 2179 | |||
XM_017018849.1 | XP_016874338.1 | 7 | 2205 | |||
GenBank mRNA | AA649108.1 | - | 1 | 137 | ||
BC032022.1 | - | 1 | 800 |