Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLIPR1L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270396.1 | NP_001257325.1 | 6 | 1479 | ||
NR_072995.1 | 7 | 1609 | ||||
XM_011537949.2 | XP_011536251.1 | 6 | 1393 | |||
XM_011537950.2 | XP_011536252.1 | 6 | 1365 | |||
XM_017018848.1 | XP_016874337.1 | 7 | 1469 | |||
XM_017018849.1 | XP_016874338.1 | 7 | 1495 | |||
GenBank mRNA | AK093175.1 | - | 6 | 1470 | ||
BC032022.1 | - | 1 | 90 | |||
BX640916.1 | - | 7 | 1578 |