Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTRK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018064.2 | NP_001018074.1 | 17 | 7654 | ||
NM_006180.4 | NP_006171.2 | 21 | 8361 | |||
XM_005252001.2 | XP_005252058.1 | 20 | 8242 | |||
XM_005252003.2 | XP_005252060.1 | 20 | 8058 | |||
XM_005252004.2 | XP_005252061.1 | 19 | 7958 | |||
XM_011518718.2 | XP_011517020.1 | 19 | 8194 | |||
XM_017014751.1 | XP_016870240.1 | 21 | 8784 | |||
XM_017014752.1 | XP_016870241.1 | 20 | 8249 | |||
XM_017014753.1 | XP_016870242.1 | 20 | 8736 | |||
XM_017014754.1 | XP_016870243.1 | 18 | 7910 | |||
GenBank mRNA | BX649001.1 | - | 1 | 3818 |