Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL13B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC073842.1 | - | 1 | 1203 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM687374.1 | - | 1 | 173 | Chr. 3 - 93981164 - 93981631 [+] on Build GRCh38 |