Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PPP2R2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177591.1 | NP_001171062.1 | 10 | 2642 | ||
NM_002717.3 | NP_002708.1 | 10 | 2910 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AL050206.1 | - | 1 | 323 | Chr. 8 - 26371375 - 26372175 [+] on Build GRCh38 |