Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYYR1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135515.1 | 4 | 3141 | |||
GenBank mRNA | AK125338.1 | - | 4 | 3141 | ||
JF693915.1 | - | 4 | 2978 |