Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUGT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011534891.2 | XP_011533193.1 | 14 | 2605 | ||
XM_011534893.2 | XP_011533195.1 | 13 | 2323 | |||
XM_017020365.1 | XP_016875854.1 | 14 | 2495 | |||
XM_017020366.1 | XP_016875855.1 | 14 | 2370 | |||
XM_017020367.1 | XP_016875856.1 | 14 | 2404 | |||
XM_017020368.1 | XP_016875857.1 | 14 | 2416 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR616065.1 | - | 1 | 1874 | Chr. 13 - 52687144 - 52689945 [+] on Build GRCh38 |