Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUC19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173600.2 | NP_775871.2 | 24870 | |||
GenBank mRNA | AK093065.1 | - | 33 | 1863 | ||
AY236870.1 | - | 32 | 1768 | |||
HM801842.1 | - | 24870 |