Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCAMP1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF086472.1 | - | 1 | 481 | ||
AK129849.1 | - | 1 | 1398 | |||
BC039455.1 | - | 2 | 1124 |