Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCAMP1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_105014.1 | 2 | 1268 | |||
GenBank mRNA | AF086472.1 | - | 1 | 430 | ||
AK129849.1 | - | 1 | 1347 | |||
BC039455.1 | - | 2 | 1073 |