Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RUNX1T1 SIRNA Assays ›
Gene Symbol:
RUNX1T1
Gene Name:
RUNX1 translocation partner 1
Gene Aliases:
AML1-MTG8, AML1T1, CBFA2T1, CDR, ETO, MTG8, ZMYND2
Chromosome Location:
Chr.8: 91954967 - 92103365 on Build GRCh38
UniGene ID:
Hs.368431
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
n331396
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RUNX1T1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001198625.1
NP_001185554.1
15
6022
NM_001198626.1
NP_001185555.1
13
5880
NM_001198627.1
NP_001185556.1
14
5940
NM_001198628.1
NP_001185557.1
12
5537
NM_001198629.1
NP_001185558.1
12
5663
NM_001198630.1
NP_001185559.1
13
5723
NM_001198631.1
NP_001185560.1
12
5736
NM_001198632.1
NP_001185561.1
12
5654
NM_001198633.1
NP_001185562.1
11
5625
NM_001198634.1
NP_001185563.1
11
5563
NM_001198679.1
NP_001185608.1
12
5707
NM_004349.3
NP_004340.1
11
5673
NM_175634.2
NP_783552.1
12
5798
NM_175635.2
NP_783553.1
11
5490
NM_175636.2
NP_783554.1
10
5572
XM_006716676.3
XP_006716739.1
13
5607
XM_011517351.2
XP_011515653.1
13
5652
XM_011517352.2
XP_011515654.1
13
5892
XM_011517353.2
XP_011515655.1
12
5600
XM_017013930.1
XP_016869419.1
12
5908
XM_017013931.1
XP_016869420.1
11
5430
XM_017013932.1
XP_016869421.1
13
5795
XM_017013933.1
XP_016869422.1
14
5859
XM_017013934.1
XP_016869423.1
14
5967
XM_017013935.1
XP_016869424.1
13
5772
XM_017013936.1
XP_016869425.1
12
5866
XM_017013937.1
XP_016869426.1
13
5692
XM_017013939.1
XP_016869428.1
7
5323
XM_017013940.1
XP_016869429.1
8
6552
XM_017013941.1
XP_016869430.1
7
6342
GenBank mRNA
AF131817.1
-
1
1233
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
Transcript Location
GenBankmRNA
BM931258.1
-
1
324
Chr. 8 - 91956510 - 91957004 [-] on Build GRCh38
GenBankmRNA
DA525131.1
-
1
159
Chr. 8 - 91956313 - 91956872 [-] on Build GRCh38
GenBankmRNA
BX431624.2
-
1
311
Chr. 8 - 91956501 - 91957023 [-] on Build GRCh38
GenBankmRNA
AI693333.1
-
1
322
Chr. 8 - 91956584 - 91957034 [-] on Build GRCh38
GenBankmRNA
CK301025.1
-
1
273
Chr. 8 - 91956512 - 91956984 [-] on Build GRCh38
Back To Top
More Information
Panther Classification:
Molecular Function -
transcription cofactor
Gene Ontology Categories:
Function(s)
Process(es)
generation of precursor metabolites and energy
transcription, DNA-templated
negative regulation of fat cell differentiation
negative regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription corepressor activity
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-qqtbl:80/10.239.164.61:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0