Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GEN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK093631.1 | - | 1 | 809 | ||
BC038243.1 | - | 1 | 1073 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR617603.1 | - | 1 | 1672 | Chr. 2 - 17753296 - 17755047 [+] on Build GRCh38 |