Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011518893.1 | XP_011517195.1 | 21 | 4896 | ||
GenBank mRNA | AF131806.1 | - | 1 | 1517 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BQ308278.1 | - | 1 | 116 | Chr. 9 - 113543877 - 113544263 [+] on Build GRCh38 |