Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRHDE-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CR936828.1 | - | 1 | 1795 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA078446.1 | - | 1 | 66 | Chr. 12 - 72255949 - 72256515 [-] on Build GRCh38 |