Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EEPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF161370.1 | - | 1 | 1153 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AF150438.1 | - | 1 | 1153 | Chr. 7 - 36281098 - 36282480 [+] on Build GRCh38 |