Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYDE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006710946.3 | XP_006711009.1 | 5 | 4182 | ||
GenBank mRNA | AK075186.1 | - | 1 | 1369 |