Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERINC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174072.2 | NP_001167543.1 | 12 | 6035 | ||
XM_011543304.1 | XP_011541606.1 | 12 | 6017 | |||
XM_017009323.1 | XP_016864812.1 | 12 | 5998 | |||
XM_017009324.1 | XP_016864813.1 | 12 | 6017 | |||
XM_017009325.1 | XP_016864814.1 | 10 | 5653 | |||
XM_017009326.1 | XP_016864815.1 | 10 | 5636 | |||
GenBank mRNA | AK026295.1 | - | 1 | 994 |