Gene Symbol: KIAA0485 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB007954.1 | - | 2 | 6480 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM759257.1 | - | 1 | 186 | Chr. 1 - 61131373 - 61131965 [+] on Build GRCh38 |