Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C7orf49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017012589.1 | XP_016868078.1 | 4 | 4405 | ||
XM_017012590.1 | XP_016868079.1 | 4 | 4331 | |||
XM_017012591.1 | XP_016868080.1 | 4 | 4185 | |||
XM_017012592.1 | XP_016868081.1 | 3 | 4366 | |||
XM_017012593.1 | XP_016868082.1 | 4 | 4235 | |||
XM_017012594.1 | XP_016868083.1 | 4 | 4442 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK056910.1 | - | 1 | 560 | Chr. 7 - 135161548 - 135163675 [-] on Build GRCh38 |