Gene Symbol: LINC01023 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126449.1 | - | 2 | 1385 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX117545.1 | - | 1 | 360 | Chr. 5 - 108726365 - 108727124 [-] on Build GRCh38 |