Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPTBN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005274192.4 | XP_005274249.1 | 38 | 8786 | ||
XM_005274193.3 | XP_005274250.1 | 37 | 8560 | |||
XM_006718669.3 | XP_006718732.1 | 36 | 10473 | |||
XM_006718671.3 | XP_006718734.1 | 38 | 8186 | |||
XM_017018174.1 | XP_016873663.1 | 38 | 8878 | |||
XM_017018175.1 | XP_016873664.1 | 38 | 8507 | |||
XM_017018176.1 | XP_016873665.1 | 38 | 8227 | |||
XM_017018177.1 | XP_016873666.1 | 37 | 8416 | |||
XM_017018178.1 | XP_016873667.1 | 37 | 8099 | |||
GenBank mRNA | AK021479.1 | - | 1 | 272 |