Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTCHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011545449.2 | XP_011543751.1 | 4 | 10542 | ||
GenBank mRNA | AK126657.1 | - | 1 | 1464 |