Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYRK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017020032.1 | XP_016875521.1 | 2 | 8422 | ||
GenBank mRNA | AK024870.1 | - | 1 | 2309 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM725582.1 | - | 1 | 334 | Chr. 12 - 67664450 - 67665108 [+] on Build GRCh38 |