Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC284244 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF070541.1 | - | 1 | 1017 | ||
AK055244.1 | - | 1 | 2434 |
Gene Symbol: ASXL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030632.2 | NP_085135.1 | 12 | 10859 | ||
XM_005258356.1 | XP_005258413.1 | 13 | 10832 | |||
XM_011526205.2 | XP_011524507.1 | 13 | 17622 | |||
XM_011526206.2 | XP_011524508.1 | 13 | 14387 | |||
XM_011526209.1 | XP_011524511.1 | 14 | 11016 | |||
XM_011526210.1 | XP_011524512.1 | 14 | 11073 | |||
XM_011526212.1 | XP_011524514.1 | 13 | 10977 | |||
XM_011526213.1 | XP_011524515.1 | 13 | 11070 | |||
XM_017026012.1 | XP_016881501.1 | 13 | 14234 | |||
XM_017026013.1 | XP_016881502.1 | 14 | 14596 | |||
XM_017026014.1 | XP_016881503.1 | 14 | 11033 |