Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100287042 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036520.1 | 1 | 1609 | |||
GenBank mRNA | AK296245.1 | - | 1 | 1539 | ||
BC021162.2 | - | 1 | 288 |