Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM120B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286379.1 | NP_001273308.1 | 11 | 4897 | ||
NM_001286380.1 | NP_001273309.1 | 11 | 4660 | |||
NM_001286381.1 | NP_001273310.1 | 9 | 2712 | |||
NM_032448.2 | NP_115824.1 | 11 | 4648 | |||
XM_017011357.1 | XP_016866846.1 | 9 | 2868 | |||
GenBank mRNA | AJ420589.1 | - | 1 | 975 |