Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ARMCX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028407.2 | 16 | 6230 | |||
NR_045861.1 | 13 | 4816 | ||||
NR_045862.1 | 13 | 4179 | ||||
NR_045864.1 | 13 | 4858 | ||||
GenBank mRNA | AL050285.1 | - | 1 | 1610 |