Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYO9A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006720539.2 | XP_006720602.1 | 43 | 11713 | ||
XM_011521613.2 | XP_011519915.1 | 44 | 11770 | |||
XM_011521614.2 | XP_011519916.1 | 45 | 11456 | |||
XM_011521615.2 | XP_011519917.1 | 44 | 11623 | |||
XM_011521616.2 | XP_011519918.1 | 44 | 11417 | |||
XM_011521617.2 | XP_011519919.1 | 44 | 11767 | |||
XM_011521618.2 | XP_011519920.1 | 43 | 11716 | |||
XM_011521619.2 | XP_011519921.1 | 43 | 11713 | |||
XM_011521620.2 | XP_011519922.1 | 43 | 11710 | |||
XM_011521621.2 | XP_011519923.1 | 43 | 11650 | |||
XM_011521622.2 | XP_011519924.1 | 43 | 11554 | |||
XM_011521623.2 | XP_011519925.1 | 27 | 8873 | |||
XM_017022228.1 | XP_016877717.1 | 44 | 11399 | |||
XM_017022229.1 | XP_016877718.1 | 44 | 11389 | |||
XM_017022230.1 | XP_016877719.1 | 42 | 11497 | |||
GenBank mRNA | BC039397.1 | - | 1 | 405 |