Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYO9A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006720539.2 | XP_006720602.1 | 43 | 12275 | ||
XM_011521613.2 | XP_011519915.1 | 44 | 12332 | |||
XM_011521614.2 | XP_011519916.1 | 45 | 12018 | |||
XM_011521615.2 | XP_011519917.1 | 44 | 12185 | |||
XM_011521616.2 | XP_011519918.1 | 44 | 11979 | |||
XM_011521617.2 | XP_011519919.1 | 44 | 12329 | |||
XM_011521618.2 | XP_011519920.1 | 43 | 12278 | |||
XM_011521619.2 | XP_011519921.1 | 43 | 12275 | |||
XM_011521620.2 | XP_011519922.1 | 43 | 12272 | |||
XM_011521621.2 | XP_011519923.1 | 43 | 12212 | |||
XM_011521622.2 | XP_011519924.1 | 43 | 12116 | |||
XM_011521623.2 | XP_011519925.1 | 27 | 9435 | |||
XM_017022228.1 | XP_016877717.1 | 44 | 11961 | |||
XM_017022229.1 | XP_016877718.1 | 44 | 11951 | |||
XM_017022230.1 | XP_016877719.1 | 42 | 12059 | |||
GenBank mRNA | BC039397.1 | - | 1 | 967 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AW972853.1 | - | 205 | Chr. - - [-] on Build |