Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101929567 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110257.1 | 6 | 7853 | |||
GenBank mRNA | BX641130.1 | - | 6 | 7860 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000454224.1 | - | 6 | 7853 | Chr. 2 - 8559833 - 8583792 [-] on Build GRCh38 |