Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCL2L13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270733.1 | NP_001257662.1 | 1 | 42 | ||
GenBank mRNA | AF086412.1 | - | 1 | 244 | ||
DC341613.1 | - | 1 | 46 |