Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC729224 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135243.1 | 3 | 1072 | |||
NR_135244.1 | 3 | 1917 | ||||
GenBank mRNA | BC040292.1 | - | 2 | 2905 |