Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCF11-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_133914.1 | 1 | 706 | |||
GenBank mRNA | AK092827.1 | - | 1 | 258 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA925269.1 | - | 1 | 259 | Chr. 11 - 83186445 - 83187036 [-] on Build GRCh38 |