Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRDM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC047115.1 | - | 1 | 476 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI668592.1 | - | 1 | 481 | Chr. 4 - 120830569 - 120831254 [-] on Build GRCh38 |