Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKAR2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321983.1 | NP_001308912.1 | 10 | 3867 | ||
NM_004157.3 | NP_004148.1 | 11 | 3933 | |||
GenBank mRNA | AK023535.1 | - | 1 | 2115 |