Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACNG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006078.3 | NP_006069.1 | 4 | 2622 | ||
XM_017028531.1 | XP_016884020.1 | 3 | 2419 | |||
GenBank mRNA | AK123431.1 | - | 1 | 146 |